Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115032.1 | NP_001108504.1 | 4 | 533 | ||
GenBank mRNA | BC158654.1 | - | 4 | 533 | 8368493 |