Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253676.1 | NP_001240605.1 | 4, 5 | 880 | ||
NM_004865.3 | NP_004856.1 | 4, 5 | 582 | |||
XM_017011513.1 | XP_016867002.1 | 6, 7 | 1155 | |||
GenBank mRNA | AB020881.1 | - | 4, 5 | 497 | ||
AF130312.1 | - | 4, 5 | 525 | |||
AF136570.1 | - | 4, 5 | 529 | |||
AK292320.1 | - | 4, 5 | 835 | |||
AK313645.1 | - | 4, 5 | 550 | |||
BC000381.2 | - | 5, 6 | 550 | 2820491 | ||
BC017559.2 | - | 4, 5 | 493 | 3911352 | ||
BI560657.1 | - | 4, 5 | 587 |