Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290229.1 | NP_001277158.1 | 6 | 768 | ||
NM_004866.5 | NP_004857.4 | 7 | 846 | |||
NR_110885.1 | 6 | 686 | ||||
GenBank mRNA | AF005037.1 | - | 7 | 720 | ||
AF038966.1 | - | 7 | 767 | |||
AK225172.1 | - | 7 | 735 | |||
AK290225.1 | - | 7 | 696 | |||
AK298146.1 | - | 5 | 479 | |||
AK304452.1 | - | 6 | 660 | |||
BC009787.1 | - | 6 | 578 | |||
BC015065.1 | - | 7 | 739 | 4105068 | ||
BC034048.1 | - | 7 | 700 | 4814951 | ||
CR542286.1 | - | 7 | 677 |