Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2v1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110345.2 | NP_001103815.2 | 4 | 1383 |
Gene Symbol: LOC100912618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751325.3 | XP_003751373.1 | 1 | 1346 |
Gene Symbol: LOC679539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751324.3 | XP_003751372.1 | 1 | 1346 |