Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290229.1 | NP_001277158.1 | 7 | 881 | ||
NM_004866.5 | NP_004857.4 | 8 | 959 | |||
NR_110885.1 | 7 | 799 | ||||
XM_011543727.2 | XP_011542029.1 | 7 | 857 | |||
GenBank mRNA | AF005037.1 | - | 8 | 833 | ||
AF038966.1 | - | 8 | 880 | |||
AK001541.1 | - | 2 | 187 | |||
AK225172.1 | - | 8 | 848 | |||
AK290225.1 | - | 8 | 809 | |||
AK298146.1 | - | 6 | 592 | |||
AK304452.1 | - | 7 | 773 | |||
BC009787.1 | - | 7 | 691 | |||
BC015065.1 | - | 8 | 852 | 4105068 | ||
BC016355.1 | - | 2 | 217 | |||
BC034048.1 | - | 8 | 813 | 4814951 | ||
CR542286.1 | - | 8 | 790 |