Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100366216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750725.3 | XP_003750773.1 | 3 | 1425 | ||
XM_006245340.2 | XP_006245402.1 | 4 | 2087 | |||
XM_006245341.1 | XP_006245403.1 | 4 | 356 | |||
XM_006245342.2 | XP_006245404.1 | 4 | 2086 | |||
XM_006245343.2 | XP_006245405.1 | 3 | 1428 | |||
XM_006245344.2 | XP_006245406.1 | 3 | 658 |
Gene Symbol: LOC103690030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245338.2 | XP_006245400.1 | 3 | 364 | ||
XM_006245339.2 | XP_006245401.1 | 3 | 838 | |||
XM_008767326.1 | XP_008765548.1 | 2 | 406 | |||
XM_008767327.1 | XP_008765549.1 | 3 | 364 |