Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109375.1 | NP_001102845.1 | 371 |
Gene Symbol: LOC103690157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773094.1 | XP_008771316.1 | 1 | 1014 |