Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190999.1 | NP_001177928.1 | 14 | 1487 | ||
XM_006227838.2 | XP_006227900.1 | 15 | 4891 | |||
XM_006227841.2 | XP_006227903.1 | 16 | 1822 | |||
XM_006227842.2 | XP_006227904.1 | 16 | 1840 | |||
XM_006227844.2 | XP_006227906.1 | 15 | 2616 | |||
XM_008758706.1 | XP_008756928.1 | 15 | 1731 | |||
XM_008758707.1 | XP_008756929.1 | 15 | 1734 |