Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190999.1 | NP_001177928.1 | 1 | 386 | ||
XM_006227838.2 | XP_006227900.1 | 2 | 3790 | |||
XM_006227841.2 | XP_006227903.1 | 3 | 721 | |||
XM_006227842.2 | XP_006227904.1 | 3 | 739 | |||
XM_006227844.2 | XP_006227906.1 | 2 | 1515 | |||
XM_008758706.1 | XP_008756928.1 | 2 | 630 | |||
XM_008758707.1 | XP_008756929.1 | 2 | 633 |