Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318876.1 | NP_001305805.1 | 8 | 955 | ||
NM_203290.3 | NP_976035.1 | 8 | 955 | |||
XM_011515000.2 | XP_011513302.1 | 8 | 896 | |||
GenBank mRNA | AF008442.1 | - | 8 | 881 | ||
AF047441.1 | - | 8 | 891 | |||
BC008118.1 | - | 8 | 846 | |||
BC008863.2 | - | 8 | 881 | 4128104 |