Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243774.2 | XP_006243836.1 | 1 | 28 | ||
XM_008766514.1 | XP_008764736.1 | 1 | 28 |