Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp90ab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004082.3 | NP_001004082.3 | 10 | 1980 | ||
GenBank mRNA | AY695392.1 | - | 10 | 1980 | ||
AY695393.1 | - | 10 | 1968 | |||
AY724481.1 | - | 10 | 1951 | |||
BC082009.1 | - | 10 | 2006 | 7131767 | ||
FQ231832.1 | - | 4 | 788 | |||
FQ233497.1 | - | 3 | 527 | |||
S45392.1 | - | 10 | 2012 |