Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126299.2 | NP_001119771.2 | 1 | 707 | ||
XM_006254412.2 | XP_006254474.1 | 3 | 1320 | |||
XM_006254413.2 | XP_006254475.1 | 3 | 1130 | |||
XM_006254414.2 | XP_006254476.1 | 3 | 1317 | |||
XM_006254415.2 | XP_006254477.1 | 3 | 1483 | |||
GenBank mRNA | BC166441.1 | - | 1 | 707 | 7461161 |