Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EI24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290135.1 | NP_001277064.1 | 10 | 1014 | ||
NM_004879.4 | NP_004870.3 | 10 | 1056 | |||
NR_110769.1 | 9 | 910 | ||||
NR_110770.1 | 11 | 1181 | ||||
XM_011543069.1 | XP_011541371.1 | 11 | 1072 | |||
XM_011543070.1 | XP_011541372.1 | 9 | 1115 | |||
XM_017018582.1 | XP_016874071.1 | 10 | 960 | |||
GenBank mRNA | AB209820.1 | - | 10 | 1013 | ||
AK223029.1 | - | 10 | 985 | |||
AK291951.1 | - | 10 | 965 | |||
AK296620.1 | - | 10 | 943 | |||
AK301556.1 | - | 9 | 839 | |||
AK315841.1 | - | 10 | 972 | |||
AK316539.1 | - | 11 | 1091 | |||
BC002390.2 | - | 10 | 922 | 2819783 | ||
BC029333.1 | - | 2 | 63 |