Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EI24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290135.1 | NP_001277064.1 | 7 | 680 | ||
NM_004879.4 | NP_004870.3 | 7 | 722 | |||
NR_110769.1 | 6 | 576 | ||||
NR_110770.1 | 8 | 847 | ||||
XM_011543069.1 | XP_011541371.1 | 8 | 738 | |||
XM_011543070.1 | XP_011541372.1 | 7 | 893 | |||
XM_017018582.1 | XP_016874071.1 | 8 | 738 | |||
GenBank mRNA | AB209820.1 | - | 7 | 679 | ||
AF010313.2 | - | 7 | 594 | |||
AK223029.1 | - | 7 | 651 | |||
AK291951.1 | - | 7 | 631 | |||
AK296620.1 | - | 7 | 609 | |||
AK301556.1 | - | 6 | 505 | |||
AK315841.1 | - | 7 | 638 | |||
AK316539.1 | - | 8 | 757 | |||
BC002390.2 | - | 7 | 588 | 2819783 |