Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126375.1 | NP_001119847.2 | 2 | 1616 | ||
XM_006244338.2 | XP_006244400.1 | 3 | 1597 | |||
GenBank mRNA | BC161872.1 | - | 2 | 1616 | 7112899 |