Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184935.1 | NP_001171864.1 | 1 | 842 | ||
NM_004883.2 | NP_004874.1 | 1 | 842 | |||
NM_013981.3 | NP_053584.1 | 1 | 842 | |||
NM_013982.2 | NP_053585.1 | 1 | 842 | |||
NM_013983.2 | NP_053586.1 | 1 | 842 | |||
XM_006714810.3 | XP_006714873.1 | 1 | 1208 | |||
XM_011537713.2 | XP_011536015.1 | 1 | 1218 | |||
XM_017010065.1 | XP_016865554.1 | 1 | 91 | |||
XM_017010066.1 | XP_016865555.1 | 1 | 1207 | |||
GenBank mRNA | AB005060.1 | - | 1 | 837 | ||
AK124504.1 | - | 2 | 771 | |||
BC103983.1 | - | 2 | 245 | |||
BC103984.1 | - | 2 | 245 | |||
BC114333.1 | - | 2 | 245 | |||
BC114334.1 | - | 2 | 245 |