Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp322a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135084.1 | NP_001128556.1 | 4 | 825 | ||
XM_006253974.1 | XP_006254036.1 | 5 | 1148 | |||
XM_006253975.1 | XP_006254037.1 | 4 | 863 | |||
XM_008771661.1 | XP_008769883.1 | 3 | 2857 | |||
XM_008771662.1 | XP_008769884.1 | 3 | 1804 | |||
XM_008771663.1 | XP_008769885.1 | 4 | 1892 | |||
XM_008771664.1 | XP_008769886.1 | 4 | 2301 | |||
XM_008771665.1 | XP_008769887.1 | 4 | 1906 | |||
XM_008771666.1 | XP_008769888.1 | 4 | 2201 | |||
XM_008771667.1 | XP_008769889.1 | 4 | 2113 | |||
XM_008771668.1 | XP_008769890.1 | 4 | 2116 | |||
XM_008771669.1 | XP_008769891.1 | 4 | 1721 | |||
XM_008771670.1 | XP_008769892.1 | 4 | 1916 | |||
GenBank mRNA | FQ209962.1 | - | 4 | 635 |