Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004283.3 | NP_004274.1 | 2 | 340 | ||
GenBank mRNA | AF081353.1 | - | 2 | 253 | ||
AF498933.1 | - | 1 | 79 | |||
AK313538.1 | - | 2 | 340 | |||
BC016471.1 | - | 2 | 317 | 3861912 | ||
DA241826.1 | - | 2 | 340 |