Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APBA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004886.3 | NP_004877.1 | 5 | 985 | ||
XM_006722950.3 | XP_006723013.1 | 5 | 1007 | |||
XM_006722951.3 | XP_006723014.1 | 3 | 692 | |||
GenBank mRNA | AB021638.1 | - | 5 | 975 | ||
AB209603.1 | - | 1 | 2214 | |||
BC008338.1 | - | 3 | 832 | |||
BC086306.1 | - | 4 | 844 | 6371716 |