Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181575.4 | NP_853553.1 | 11 | 1450 | ||
NR_126510.1 | 11 | 1512 | ||||
NR_126511.1 | 10 | 1646 | ||||
XM_005264392.3 | XP_005264449.1 | 10 | 1395 | |||
GenBank mRNA | AF100746.1 | - | 11 | 1223 | ||
AF100753.1 | - | 11 | 1250 | |||
AF100754.1 | - | 10 | 1448 | |||
AF165515.1 | - | 11 | 1230 | |||
AK023983.1 | - | 11 | 1290 | |||
AK055566.1 | - | 7 | 2082 | |||
AK293121.1 | - | 11 | 1269 | |||
AK304886.1 | - | 10 | 1392 | |||
BC001658.1 | - | 11 | 1375 | 3050204 | ||
BC033646.2 | - | 11 | 1265 | |||
CR749661.1 | - | 10 | 1383 |