Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249208.2 | XP_006249270.1 | 2 | 356 | ||
XM_341056.7 | XP_341057.3 | 2 | 356 |