Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127502.1 | NP_001120974.1 | 3 | 516 | ||
XM_006257281.2 | XP_006257343.1 | 3 | 555 | |||
XM_008773438.1 | XP_008771660.1 | 3 | 536 | |||
GenBank mRNA | BC166897.1 | - | 3 | 516 | 6921863 | |
FQ215245.1 | - | 3 | 444 | |||
FQ220910.1 | - | 3 | 498 | |||
FQ232216.1 | - | 3 | 490 |