Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC680370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109402.1 | NP_001102872.1 | 2 | 484 | ||
XM_008773231.1 | XP_008771453.1 | 3 | 551 | |||
XM_008773232.1 | XP_008771454.1 | 4 | 594 |