Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181575.4 | NP_853553.1 | 3 | 578 | ||
NR_126510.1 | 3 | 578 | ||||
NR_126511.1 | 3 | 578 | ||||
XM_005264392.3 | XP_005264449.1 | 2 | 523 | |||
GenBank mRNA | AF100746.1 | - | 3 | 351 | ||
AF100753.1 | - | 3 | 378 | |||
AF100754.1 | - | 3 | 378 | |||
AF165515.1 | - | 3 | 358 | |||
AK023983.1 | - | 3 | 356 | |||
AK055566.1 | - | 2 | 477 | |||
AK293121.1 | - | 3 | 397 | |||
AK304886.1 | - | 2 | 520 | |||
AK308231.1 | - | 3 | 379 | |||
AK310159.1 | - | 3 | 379 | |||
BC001658.1 | - | 3 | 441 | 3050204 | ||
BC033646.2 | - | 3 | 393 | |||
CR749661.1 | - | 2 | 449 | |||
CR999537.1 | - | 3 | 564 | |||
CX164419.1 | - | 3 | 360 |