Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004890.2 | NP_004881.2 | 7 | 646 | ||
XM_006721600.2 | XP_006721663.1 | 7 | 1271 | |||
GenBank mRNA | AF047437.1 | - | 7 | 623 | ||
AK123065.1 | - | 1425 | ||||
AK312174.1 | - | 7 | 622 | |||
BC011934.2 | - | 7 | 615 | 4330461 |