Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dqx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109407.1 | NP_001102877.1 | 11 | 2013 | ||
XM_006236717.2 | XP_006236779.1 | 11 | 2503 | |||
XM_008763030.1 | XP_008761252.1 | 10 | 2688 |
Gene Symbol: LOC103692182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763119.1 | XP_008761341.1 | 569 |