Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004891.3 | NP_004882.1 | 4 | 262 | ||
NM_145330.2 | NP_663303.1 | 3 | 155 | |||
GenBank mRNA | AF047440.1 | - | 4 | 236 | ||
BC009475.1 | - | 4 | 214 | 4049053 | ||
BC020834.1 | - | 4 | 249 | 4775045 | ||
BC087846.1 | - | 3 | 119 | |||
BC090047.1 | - | 3 | 155 | 30393466 | ||
BI601959.1 | - | 4 | 265 |