Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS26A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035260.2 | NP_001030337.1 | 6 | 755 | ||
NM_001318944.1 | NP_001305873.1 | 7 | 861 | |||
NM_001318945.1 | NP_001305874.1 | 7 | 831 | |||
NM_001318946.1 | NP_001305875.1 | 5 | 679 | |||
NM_004896.4 | NP_004887.2 | 6 | 755 | |||
GenBank mRNA | AF054179.1 | - | 6 | 709 | ||
AF175266.1 | - | 6 | 709 | |||
AK022992.1 | - | 6 | 685 | |||
AK290536.1 | - | 5 | 585 | |||
AK309021.1 | - | 7 | 809 | |||
BC022505.1 | - | 6 | 742 | 4799288 | ||
BQ048905.1 | - | 5 | 624 | |||
BX647725.1 | - | 5 | 600 | |||
BX649092.1 | - | 5 | 654 |