Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207007.3 | NP_996890.1 | 3 | 451 | ||
NR_111969.1 | 2 | 336 | ||||
GenBank mRNA | AY766447.1 | - | 3 | 296 | ||
AY766449.1 | - | 2 | 181 | |||
BC130456.1 | - | 3 | 258 | 40146487 | ||
BC130458.1 | - | 3 | 258 | 40146489 | ||
BC144393.1 | - | 3 | 258 | 9052914 | ||
BC144394.1 | - | 3 | 258 | 9052915 | ||
BC146944.1 | - | 3 | 451 | 9056872 | ||
BC146958.1 | - | 3 | 451 | 9056886 | ||
BC171857.1 | - | 3 | 451 | 9054511 | ||
BC171864.1 | - | 3 | 451 | 9054518 |
Gene Symbol: CCL4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291468.1 | NP_001278397.1 | 2, 3 | 281 | ||
NM_001291469.1 | NP_001278398.1 | 3 | 485 | |||
NM_001291470.1 | NP_001278399.1 | 2 | 887 | |||
NM_001291471.1 | NP_001278400.1 | 3 | 465 | |||
NM_001291472.1 | NP_001278401.1 | 3 | 444 | |||
NM_001291473.1 | NP_001278402.1 | 3 | 414 | |||
NM_001291474.1 | NP_001278403.1 | 2 | 370 | |||
NM_001291475.1 | NP_001278404.1 | 3 | 475 | |||
NR_111970.1 | 1, 2 | 166 | ||||
GenBank mRNA | AY766450.1 | - | 2, 3 | 281 | ||
AY766451.1 | - | 3 | 485 | |||
AY766452.1 | - | 2 | 887 | |||
AY766453.1 | - | 3 | 465 | |||
AY766454.1 | - | 3 | 444 | |||
AY766455.1 | - | 3 | 414 | |||
AY766456.1 | - | 1, 2 | 166 | |||
AY766457.1 | - | 2 | 370 | |||
AY766458.1 | - | 3 | 475 | |||
BC092445.1 | - | 3 | 482 | 30416435 | ||
CD521054.1 | - | 1 | 214 |