Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou5f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081751.1 | NP_001075220.1 | 1 | 705 | ||
GenBank mRNA | L23864.1 | - | 1 | 705 |