Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam19a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765429.1 | XP_008763651.1 | 3, 4 | 1068 | ||
XM_008765430.1 | XP_008763652.1 | 3, 4 | 1902 |