Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017187.2 | NP_058883.1 | 4 | 597 | ||
GenBank mRNA | BC078866.1 | - | 3 | 455 | 7114119 | |
BC089854.1 | - | 4 | 597 | 7380208 | ||
BC107455.1 | - | 4 | 626 | 7377505 | ||
D84418.1 | - | 4 | 519 | |||
FQ225084.1 | - | 4 | 561 | |||
FQ229618.1 | - | 4 | 534 | |||
FQ230440.1 | - | 4 | 561 | |||
FQ233747.1 | - | 4 | 535 |
Gene Symbol: Hmgb2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_573272.5 | XP_573272.3 | 1 | 534 |
Gene Symbol: RGD1559962 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758645.1 | XP_008756867.1 | 1 | 1949 |