Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004286.4 | NP_004277.2 | 11 | 1930 | ||
XM_011530537.2 | XP_011528839.1 | 11 | 3192 | |||
XM_017029099.1 | XP_016884588.1 | 11 | 3184 | |||
XM_017029100.1 | XP_016884589.1 | 11 | 3154 | |||
XM_017029101.1 | XP_016884590.1 | 12 | 1771 | |||
XM_017029102.1 | XP_016884591.1 | 10 | 1587 | |||
XM_017029103.1 | XP_016884592.1 | 10 | 1685 | |||
GenBank mRNA | AK096647.1 | - | 1 | 1266 | ||
AK315704.1 | - | 10 | 1553 | |||
BC014075.1 | - | 11 | 1768 | |||
CR456501.1 | - | 11 | 1753 | |||
U87964.1 | - | 11 | 1674 |