Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Treml4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244488.2 | XP_006244550.1 | 1 | 683 | ||
XM_006244490.2 | XP_006244552.1 | 1 | 687 | |||
XM_006244492.2 | XP_006244554.1 | 1 | 692 |