Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191075.1 | NP_001178004.1 | 7 | 628 | ||
XM_006247427.2 | XP_006247489.1 | 6 | 651 |