Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191077.1 | NP_001178006.1 | 8 | 997 | ||
XM_006232755.2 | XP_006232817.1 | 8 | 1999 | |||
XM_008761210.1 | XP_008759432.1 | 7 | 2074 | |||
XM_008761211.1 | XP_008759433.1 | 7 | 1942 | |||
XM_008761212.1 | XP_008759434.1 | 7 | 1179 | |||
XM_008761213.1 | XP_008759435.1 | 6 | 1669 |
Set Membership: |
G-protein coupled receptor |