Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253854.1 | NP_001240783.1 | 11 | 1569 | ||
NM_001253855.1 | NP_001240784.1 | 7 | 1300 | |||
NM_012443.3 | NP_036575.1 | 8 | 1214 | |||
NM_172242.2 | NP_758442.1 | 8 | 1214 | |||
XM_005252645.1 | XP_005252702.1 | 8 | 1243 | |||
XM_005252646.1 | XP_005252703.1 | 7 | 1069 | |||
GenBank mRNA | AF079363.1 | - | 8 | 1171 | ||
AK289903.1 | - | 8 | 1163 | |||
AK302194.1 | - | 11 | 1544 | |||
AK310247.1 | - | 7 | 1253 | |||
AL080136.1 | - | 8 | 1162 | |||
BC030585.2 | - | 8 | 1162 | 4822996 | ||
CR533552.1 | - | 8 | 1047 | |||
HM370395.1 | - | 3 | 225 |