Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E4f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246025.2 | XP_006246087.1 | 15 | 5116 | ||
XM_006246026.2 | XP_006246088.1 | 14 | 5203 | |||
XM_008767590.1 | XP_008765812.1 | 14 | 5200 | |||
XM_008767591.1 | XP_008765813.1 | 14 | 4807 |
Gene Symbol: Dnase1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001057412.3 | XP_001057412.4 | 4 | 557 | ||
XM_006246041.1 | XP_006246103.1 | 4 | 557 | |||
XM_006246042.1 | XP_006246104.1 | 4 | 557 |