Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVPR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000707.3 | NP_000698.1 | 2 | 1173 | ||
GenBank mRNA | AF030512.1 | - | 2 | 1138 | ||
AF101726.1 | - | 2 | 1138 | |||
D31833.1 | - | 2 | 1222 | |||
EU432111.1 | - | 2 | 1015 | |||
L37112.1 | - | 2 | 1109 |
Set Membership: |
G-protein coupled receptor |