Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284259.1 | NP_001271188.1 | 31 | 5241 | ||
NM_016195.3 | NP_057279.2 | 31 | 5121 | |||
GenBank mRNA | AB033337.1 | - | 31 | 5194 | ||
AK025628.1 | - | 11 | 1276 | |||
AL117496.1 | - | 31 | 5076 | |||
AL137392.1 | - | 18 | 3708 | |||
AY282407.1 | - | 16 | 3038 | |||
AY739715.1 | - | 13 | 2140 | |||
L16782.1 | - | 12 | 1388 | |||
U93121.1 | - | 12 | 1949 |