Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284259.1 | NP_001271188.1 | 20 | 3211 | ||
NM_016195.3 | NP_057279.2 | 20 | 3091 | |||
GenBank mRNA | AB033337.1 | - | 20 | 3164 | ||
AK024959.1 | - | 3 | 786 | |||
AL117496.1 | - | 20 | 3046 | |||
AL137392.1 | - | 7 | 1678 | |||
AY282407.1 | - | 5 | 1008 |