Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284259.1 | NP_001271188.1 | 28 | 4807 | ||
NM_016195.3 | NP_057279.2 | 28 | 4687 | |||
GenBank mRNA | AB033337.1 | - | 28 | 4760 | ||
AK025628.1 | - | 8 | 842 | |||
AL117496.1 | - | 28 | 4642 | |||
AL137392.1 | - | 15 | 3274 | |||
AY282407.1 | - | 13 | 2604 | |||
AY739715.1 | - | 9 | 1400 | |||
L16782.1 | - | 9 | 954 | |||
U93121.1 | - | 9 | 1515 |