Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAD2L1BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003690.1 | NP_001003690.1 | 3 | 467 | ||
NM_014628.2 | NP_055443.1 | 2 | 199 | |||
GenBank mRNA | AK297172.1 | - | 2 | 220 | ||
AL537692.3 | - | 3 | 469 | |||
BC002904.2 | - | 2 | 169 | 3945348 | ||
BE297042.1 | - | 3 | 395 | |||
BI255998.1 | - | 2 | 186 | |||
BU845738.1 | - | 3 | 504 | |||
CB991410.1 | - | 2 | 226 | |||
CR456893.1 | - | 2 | 163 | |||
D14811.1 | - | 2 | 166 |