Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAD2L1BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003690.1 | NP_001003690.1 | 4 | 966 | ||
NM_014628.2 | NP_055443.1 | 3 | 698 | |||
GenBank mRNA | AK297172.1 | - | 3 | 719 | ||
BC002904.2 | - | 3 | 668 | 3945348 | ||
BI255998.1 | - | 3 | 686 | |||
CB991410.1 | - | 3 | 725 | |||
CR456893.1 | - | 3 | 662 | |||
D14811.1 | - | 3 | 665 |