Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps27l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276477.1 | NP_001263406.1 | 3 | 366 | ||
GenBank mRNA | CB712651.1 | - | 2 | 116 | ||
FQ109437.1 | - | 3 | 365 | |||
FQ209888.1 | - | 2 | 163 | |||
X59375.1 | - | 2 | 164 |