Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100362173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002730241.3 | XP_002730287.1 | 3 | 810 | ||
XM_006256957.2 | XP_006257019.1 | 3 | 735 |
Gene Symbol: LOC680342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001056750.4 | XP_001056750.2 | 3 | 801 |