Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004288.4 | NP_004279.3 | 1 | 202 | ||
GenBank mRNA | AF068836.1 | - | 1 | 163 | ||
AK312753.1 | - | 1 | 199 | |||
BC036449.2 | - | 1 | 160 | 4830272 | ||
CD642163.1 | - | 1 | 255 | |||
DB219691.1 | - | 1 | 203 | |||
L06633.1 | - | 1 | 116 |