Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004288.4 | NP_004279.3 | 8 | 1095 | ||
XM_017005386.1 | XP_016860875.1 | 8 | 1286 | |||
GenBank mRNA | AF068836.1 | - | 8 | 1056 | ||
AK301544.1 | - | 11 | 1188 | |||
AK312753.1 | - | 8 | 1092 | |||
BC036449.2 | - | 8 | 1053 | 4830272 |