Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004911.4 | NP_004902.1 | 9 | 1786 | ||
XM_006716185.2 | XP_006716248.1 | 9 | 1789 | |||
GenBank mRNA | AK290971.1 | - | 9 | 1571 | ||
AK313299.1 | - | 9 | 1758 | |||
BC000425.2 | - | 9 | 1548 | 2819726 | ||
BC001928.1 | - | 9 | 1548 | 2819726 | ||
BC006344.2 | - | 9 | 1736 | 4098367 | ||
BC011754.2 | - | 9 | 1737 | 3532913 |